Anti-Cytomegalovirus Late Nuclear Antigen Mouse monoclonal antibody [clone: BM219]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9099-04KEA 3620 DKK
USBIC9099-04K
Anti-Cytomegalovirus Late Nuclear Antigen Mouse monoclonal antibody [clone: BM219]
Antistoffer
Recommended dilution: ELISA: 1:500-1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: Cytomegalovirus Late Nuclear Antigen
Clonality: Monoclonal
Clone: BM219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR