Anti-Cytomegalovirus Late Nuclear Antigen Mouse monoclonal antibody [clone: 10B2679]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9099-04JEA 4880 DKK
USBIC9099-04J
Anti-Cytomegalovirus Late Nuclear Antigen Mouse monoclonal antibody [clone: 10B2679]
Antistoffer
  • Recommended dilution: Immunofluorescence: 0. 02-2μg/slide

Type: Primary
Antigen: Cytomegalovirus Late Nuclear Antigen
Clonality: Monoclonal
Clone: 10B2679
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR