Anti-Cytomegalovirus Immediate/Early Antigen Mouse monoclonal antibody [clone: 10B2671]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9098-46EA 5120 DKK
USBIC9098-46
Anti-Cytomegalovirus Immediate/Early Antigen Mouse monoclonal antibody [clone: 10B2671]
Antistoffer
Type: Primary
Antigen: Cytomegalovirus Immediate/Early Antigen
Clonality: Monoclonal
Clone: 10B2671
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR