Anti-Cytokeratin Type II Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F224]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-61FEA 3730 DKK
USBIC9097-61F
Anti-Cytokeratin Type II Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F224]
Antistoffer
Recommended dilution: Immunofluorescence: 1:20-1:40

Type: Primary
Antigen: Cytokeratin Type II
Clonality: Monoclonal
Clone: 5F224
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR