Anti-Cytokeratin, pan Mouse monoclonal antibody [clone: 10K183]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-45AEA 3290 DKK
USBIC9097-45A
Anti-Cytokeratin, pan Mouse monoclonal antibody [clone: 10K183]
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2-5 ?g/ml

Type: Primary
Antigen: Cytokeratin, pan
Clonality: Monoclonal
Clone: 10K183
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a+IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR