Anti-KRT8 Mouse Monoclonal Antibody (Biotin) [clone: 5F235 (Ks8.7)]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-42J1EA 3870 DKK
USBIC9097-42J1
Anti-KRT8 Mouse Monoclonal Antibody (Biotin) [clone: 5F235 (Ks8.7)]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: Monoclonal
Clone: 5F235 (Ks8.7)
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR