Anti-KRT8 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-42B2EA 2590 DKK
USBIC9097-42B2
Anti-KRT8 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR