Anti-Cytokeratin 7,17,19 Mouse monoclonal antibody [clone: 4F5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-40ZEA 5200 DKK
USBIC9097-40Z
Anti-Cytokeratin 7,17,19 Mouse monoclonal antibody [clone: 4F5]
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Cytokeratin 7,17,19
Clonality: Monoclonal
Clone: 4F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR