Anti-KRT7 Mouse Monoclonal Antibody [clone: 10g30]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-40REA 3700 DKK
USBIC9097-40R
Anti-KRT7 Mouse Monoclonal Antibody [clone: 10g30]
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2.0-4.0 ?g/ml

Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: Monoclonal
Clone: 10g30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR