Anti-KRT7 Mouse Monoclonal Antibody [clone: 5F234]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-39DEA 3810 DKK
USBIC9097-39D
Anti-KRT7 Mouse Monoclonal Antibody [clone: 5F234]
Antistoffer
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: Monoclonal
Clone: 5F234
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig, Sheep
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR