Anti-KRT7 Mouse Monoclonal Antibody [clone: 5F282]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC9097-39A1EA 4440 DKK
USBIC9097-39A1
Anti-KRT7 Mouse Monoclonal Antibody [clone: 5F282]
Antistoffer
Recommended dilution: Immunochemistry: 1:5-1:10
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: Monoclonal
Clone: 5F282
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Human, Mouse