Anti-Cytokeratin 6 Mouse Monoclonal Antibody [clone: 5F222]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-38DEA 4290 DKK
USBIC9097-38D
Anti-Cytokeratin 6 Mouse Monoclonal Antibody [clone: 5F222]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Cytokeratin 6
Clonality: Monoclonal
Clone: 5F222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR