Anti-Cytokeratin 5 + 14 Mouse Monoclonal Antibody [clone: 8J1213]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-37REA 6350 DKK
USBIC9097-37R
Anti-Cytokeratin 5 + 14 Mouse Monoclonal Antibody [clone: 8J1213]
Antistoffer
Type: Primary
Antigen: Cytokeratin 5 + 14
Clonality: Monoclonal
Clone: 8J1213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG3
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR