Anti-KRT5 Mouse Monoclonal Antibody [clone: 1E1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-37N2EA 4320 DKK
USBIC9097-37N2
Anti-KRT5 Mouse Monoclonal Antibody [clone: 1E1]
Antistoffer
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:400

Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: Monoclonal
Clone: 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR