Anti-Cytokeratin 5,8 Mouse Monoclonal Antibody [clone: 5F173]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-37KEA 4260 DKK
USBIC9097-37K
Anti-Cytokeratin 5,8 Mouse Monoclonal Antibody [clone: 5F173]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Cytokeratin 5,8
Clonality: Monoclonal
Clone: 5F173
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR