Anti-KRT4 Mouse monoclonal antibody Biotin [clone: 10B2649]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-35G-BIOTEA 0 DKK
USBIC9097-35G-BIOT
Anti-KRT4 Mouse monoclonal antibody Biotin [clone: 10B2649]
Type: Primary
Antigen: KRT4 (keratin 4)
Clonality: Monoclonal
Clone: 10B2649
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR