Anti-Cytokeratin 2e Mouse Monoclonal Antibody [clone: 5D49]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-33EA 3450 DKK
USBIC9097-33
Anti-Cytokeratin 2e Mouse Monoclonal Antibody [clone: 5D49]
Antistoffer
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: Cytokeratin 2e
Clonality: Monoclonal
Clone: 5D49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR