Anti-KRT20 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29PEA 3810 DKK
USBIC9097-29P
Anti-KRT20 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR