Anti-KRT20 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2648]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29G-APCEA 0 DKK
USBIC9097-29G-APC
Anti-KRT20 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2648]
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Monoclonal
Clone: 10B2648
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR