Anti-KRT20 Mouse Monoclonal Antibody [clone: 10B2648]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29GEA 3500 DKK
USBIC9097-29G
Anti-KRT20 Mouse Monoclonal Antibody [clone: 10B2648]
Antistoffer
Recommended dilution: Immunofluorescence: 10 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Monoclonal
Clone: 10B2648
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR