Anti-KRT20 Mouse Monoclonal Antibody [clone: 6A60]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29EEA 3640 DKK
USBIC9097-29E
Anti-KRT20 Mouse Monoclonal Antibody [clone: 6A60]
Antistoffer
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Monoclonal
Clone: 6A60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR