Anti-KRT20 Mouse Monoclonal Antibody [clone: 5F219]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29DEA 5430 DKK
USBIC9097-29D
Anti-KRT20 Mouse Monoclonal Antibody [clone: 5F219]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Monoclonal
Clone: 5F219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR