Anti-KRT20 Mouse Monoclonal Antibody [clone: 5F215]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-29EA 5060 DKK
USBIC9097-29
Anti-KRT20 Mouse Monoclonal Antibody [clone: 5F215]
Antistoffer
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: Monoclonal
Clone: 5F215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR