Anti-KRT19 Mouse Monoclonal Antibody [clone: 5F230]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC9097-28EA 4590 DKK
USBIC9097-28
Anti-KRT19 Mouse Monoclonal Antibody [clone: 5F230]
Antistoffer
Recommended dilution: Immunochemistry: 1:10
Type: Primary
Antigen: KRT19 (keratin 19)
Clonality: Monoclonal
Clone: 5F230
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Rabbit, Rat