Anti-KRT19 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-24CEA 4320 DKK
USBIC9097-24C
Anti-KRT19 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml; ELISA: 1:1000

Type: Primary
Antigen: KRT19 (keratin 19)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR