Anti-KRT18 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2644]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-23F-FITCEA 0 DKK
USBIC9097-23F-FITC
Anti-KRT18 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2644]
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone: 10B2644
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR