Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: 5F227]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-23D2EA 5140 DKK
USBIC9097-23D2
Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: 5F227]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone: 5F227
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Hamster, Human, Mouse, Pig, Rat, Sheep
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR