Anti-KRT18 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-21NEA 4320 DKK
USBIC9097-21N
Anti-KRT18 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:25-200

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR