Anti-KRT18 Mouse Monoclonal Antibody [clone: 5F301]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-21A1EA 4920 DKK
USBIC9097-21A1
Anti-KRT18 Mouse Monoclonal Antibody [clone: 5F301]
Antistoffer
Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone: 5F301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Dog, Human, Mammalian, Mouse, Pig, Rabbit, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR