Anti-KRT17 Guinea pig Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-17AEA 4510 DKK
USBIC9097-17A
Anti-KRT17 Guinea pig Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:5000-1:10000

Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR