Anti-KRT17 Mouse monoclonal antibody [clone: 10B2642]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-16JEA 0 DKK
USBIC9097-16J
Anti-KRT17 Mouse monoclonal antibody [clone: 10B2642]
Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: Monoclonal
Clone: 10B2642
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR