Anti-KRT15 Guinea pig Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-15AEA 4900 DKK
USBIC9097-15A
Anti-KRT15 Guinea pig Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: KRT15 (keratin 15)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR