Anti-KRT14 Guinea pig Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC9097-13GEA 5440 DKK
USBIC9097-13G
Anti-KRT14 Guinea pig Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:5000-1:10000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: KRT14 (keratin 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Bovine, Human, Mouse