Anti-KRT14 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3H2053]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-13F1EA 4190 DKK
USBIC9097-13F1
Anti-KRT14 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3H2053]
Antistoffer
Type: Primary
Antigen: KRT14 (keratin 14)
Clonality: Monoclonal
Clone: 3H2053
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR