Anti-KRT13 Mouse Monoclonal Antibody [clone: 5F83]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-12DEA 3460 DKK
USBIC9097-12D
Anti-KRT13 Mouse Monoclonal Antibody [clone: 5F83]
Antistoffer
Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: KRT13 (keratin 13)
Clonality: Monoclonal
Clone: 5F83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mammalian, Mouse, Rabbit, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR