Anti-KRT10 Mouse Monoclonal Antibody [clone: 5F187]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-09BEA 4640 DKK
USBIC9097-09B
Anti-KRT10 Mouse Monoclonal Antibody [clone: 5F187]
Antistoffer
Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: KRT10 (keratin 10)
Clonality: Monoclonal
Clone: 5F187
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammalian
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR