Anti-Cytokeratin Mouse monoclonal antibody [clone: 10B2635]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9097-01XEA 6730 DKK
USBIC9097-01X
Anti-Cytokeratin Mouse monoclonal antibody [clone: 10B2635]
Antistoffer
Recommended dilution: Immunochemistry: 1:150-1:750

Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: 10B2635
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Rat, Xenopus laevis
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR