Anti-CYTH2 Mouse monoclonal antibody Biotin [clone: 11C278]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9096-10D-BIOTEA 0 DKK
USBIC9096-10D-BIOT
Anti-CYTH2 Mouse monoclonal antibody Biotin [clone: 11C278]
Type: Primary
Antigen: CYTH2 (cytohesin 2)
Clonality: Monoclonal
Clone: 11C278
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR