Anti-CYTH2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9096-10CEA 4690 DKK
USBIC9096-10C
Anti-CYTH2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: CYTH2 (cytohesin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR