Anti-CYTH1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 11C277]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9096-05E-FITCEA 0 DKK
USBIC9096-05E-FITC
Anti-CYTH1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 11C277]
Type: Primary
Antigen: CYTH1 (cytohesin 1)
Clonality: Monoclonal
Clone: 11C277
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR