Anti-CYTH1 Rat Monoclonal Antibody [clone: 10H4]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC9096-05CEA 4590 DKK
USBIC9096-05C
Anti-CYTH1 Rat Monoclonal Antibody [clone: 10H4]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: CYTH1 (cytohesin 1)
Clonality: Monoclonal
Clone: 10H4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Human