Anti-CYP4F3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-45BEA 4380 DKK
USBIC9095-45B
Anti-CYP4F3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: CYP4F3 (cytochrome P450, family 4, subfamily F, polypeptide 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR