Anti-CYP19A1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-19X1EA 3460 DKK
USBIC9095-19X1
Anti-CYP19A1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR