Anti-CYP19A1 Mouse Monoclonal Antibody [clone: 10B2634]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-19W2EA 7120 DKK
USBIC9095-19W2
Anti-CYP19A1 Mouse Monoclonal Antibody [clone: 10B2634]
Antistoffer
Recommended dilution: Western Blot: 1:250

Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: Monoclonal
Clone: 10B2634
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Chicken, Goat, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR