Anti-Cytochrome P450 4A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-16EA 4090 DKK
USBIC9095-16
Anti-Cytochrome P450 4A Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:100; Western Blot: 1:500

Type: Primary
Antigen: Cytochrome P450 4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cat, Rabbit, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR