Anti-Cytochrome P450 3A4 Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC9095-15N2EA 7050 DKK
USBIC9095-15N2
Anti-Cytochrome P450 3A4 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:2000
Type: Primary
Antigen: Cytochrome P450 3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat