Anti-CYP2E1 Mouse Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-15M5EA 3500 DKK
USBIC9095-15M5
Anti-CYP2E1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP2E1 (cytochrome P450, family 2, subfamily E, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR