Anti-CYP2D6 Mouse Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-15K4EA 3500 DKK
USBIC9095-15K4
Anti-CYP2D6 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP2D6 (cytochrome P450, family 2, subfamily D, polypeptide 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR