Anti-CYP2C9 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-15J2EA 4380 DKK
USBIC9095-15J2
Anti-CYP2C9 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CYP2C9 (cytochrome P450, family 2, subfamily C, polypeptide 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR