Anti-CYP46A1 Mouse Monoclonal Antibody [clone: 9G345]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-14DEA 4490 DKK
USBIC9095-14D
Anti-CYP46A1 Mouse Monoclonal Antibody [clone: 9G345]
Antistoffer
Type: Primary
Antigen: CYP46A1 (cytochrome P450, family 46, subfamily A, polypeptide 1)
Clonality: Monoclonal
Clone: 9G345
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR